MilliporeSigma
  • Blood kinetics of sulfamonomethoxine and oxytetracycline following intrauterine spray injection in dairy cows.
Blood kinetics of sulfamonomethoxine and oxytetracycline following intrauterine spray injection in dairy cows.
Pharmacological research (1990-03-01)
C Girardi, G Re, A M Farca, M Dacasto, E Ferrero, M Eandi
ABSTRACT
Intrauterine administration was performed in six Friesan cows with a disposable spray preparation containing 3 g of sulfamonomethoxine and 3 g of oxytetracycline, in order to investigate their serum kinetics. Sulfamonomethoxine levels were determined by a reversed-phase HPLC method, whilst oxytetracycline quantities were detected by a microbiological method. The sulphonamide had a peak 1.17 h after the administration, the tetracycline reached its highest concentration after 8 h. The bioavailability of both drugs was low and detectable drug amounts were no longer recovered after 24 h.
Sigma-Aldrich
Sulfamonomethoxine