MilliporeSigma
Topical delivery of siRNA into skin using ionic liquids.
Journal of controlled release : official journal of the Controlled Release Society (2020-04-28)
Vimisha Dharamdasani, Abhirup Mandal, Qin M Qi, Isabella Suzuki, Maria Vitória Lopes Badra Bentley, Samir Mitragotri
ABSTRACT
Skin diseases such as lupus, cancer, psoriasis, and hyperhidrosis can potentially be treated effectively by suppressing allele-specific genes using small interfering RNA (siRNA). Injections of siRNA into skin, though effective, are painful and cover small surface areas and thus are not suitable as a long-term treatment option. Topical delivery of siRNA is an attractive alternative option to mediate RNA interference (RNAi). However, the barrier function of the epidermis impedes effective permeation of siRNA into the skin. Herein, we describe topical delivery of siRNA using ionic liquids (ILs) capable of complexing with siRNA non-covalently and delivering it effectively. Using complementary and synergistic strategies of ionic liquids, we report delivery of effective doses of siRNA into skin. The first strategy involved the use of hydrophobic cations to robe the siRNA and the second strategy involved the use of choline-geranic acid ionic liquid (CAGE) to enhance its dermal penetration. In vitro studies in porcine skin confirmed the synergistic effect of these strategies in enhancing epidermal and dermal penetration. In vivo application of siRNA formulation to SKH-1E hairless mice significantly suppressed GAPDH expression with no clinical evidence of toxicity. This is a simple, personalized, and scalable platform for effective topical delivery of siRNA for treating genetic skin diseases.
Sigma-Aldrich
Choline bicarbonate, ~80% in H2O