MilliporeSigma
  • Effects of frequently applied carbon monoxide releasing molecules (CORMs) in typical CO-sensitive model systems - A comparative in vitro study.
Effects of frequently applied carbon monoxide releasing molecules (CORMs) in typical CO-sensitive model systems - A comparative in vitro study.
Archives of biochemistry and biophysics (2020-04-27)
David Stucki, Heide Krahl, Moritz Walter, Julia Steinhausen, Katrin Hommel, Peter Brenneisen, Wilhelm Stahl
ABSTRACT
Intracellular carbon monoxide (CO) is a gaseous signaling molecule and is generated enzymatically by heme oxygenases upon degradation of heme to billiverdin. Target structures for intracellular produced CO are heme proteins including cytochrome c oxidase of the respiratory chain, cytochrome P450-dependent monooxygenases, or myoglobin. For studies on CO signaling, CO-releasing molecules (CORMs) of different structure are available. Here, three frequently used CORMs (CORM-2, CORM-3 and CORM-401) were studied for their properties to provide CO in biological test systems and address susceptible heme proteins. CO release was investigated in the myoglobin binding assay and found to be rapid (<5 min) with CORM-2- and CORM-3, whereas CORM-401 continuously provided CO (>50 min). Storage stability of CORM stock solutions was also assessed with the myoglobin assay. Only CORM-401 stock solutions were stable over a period of 7 days. Incubation of CORMs with recombinant cytochrome P450 led to an inhibition of enzyme activity. However, only CORM-3 and CORM-401 proved to be suitable in this test system because controls with the inactivated CORM-2 (iCORM-2) also led to a loss of enzyme activity. The impact of CORMs on the respiratory chain was investigated with high resolution respirometry and extracellular flux technology. In the first approach interferences of CORM-2 and CORM-3 with oxygen measurement occurred, since a rapid depletion of oxygen was detected in the medium even when no cells were present. However, CORM-401 did not interfere with oxygen measurement and the expected inhibition of cellular respiration was observed. CORM-2 was not suitable for use in oxygen measurements with the extracellular flux technology and CORM-3 application did not show any effect in this system. However, CO-dependent inhibition of cellular respiration was observed with CORM-401. Based on the present experiments it is concluded, that CORM-401 produced most reliable CO-specific results for the modulation of typical CO targets. For studies on CO-dependent biological effects on intracellular heme groups, CORM-2 and CORM-3 were less suitable. Depending on the experimental setting, data achieved with these compounds should be evaluated with caution.
Sigma-Aldrich
Tricarbonyldichlororuthenium(II) dimer
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Cytochrome P450 human, 1A1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)