MilliporeSigma
  • Determination of four nitrofuran metabolites in gelatin Chinese medicine using dispersive solid phase extraction and pass-through solid phase extraction coupled to ultra high performance liquid chromatography-tandem mass spectrometry.
Determination of four nitrofuran metabolites in gelatin Chinese medicine using dispersive solid phase extraction and pass-through solid phase extraction coupled to ultra high performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2020-04-26)
Jinyan Gong, Jiong Li, Haina Yuan, Bingquan Chu, Weijie Lin, Qingwen Cao, Qiqi Zhao, Ruosi Fang, Ling Li, Gongnian Xiao
ABSTRACT
This study established a validated analytical method for the first time on the determination of nitrofuran metabolites, including semicarbazide (SEM), 1-aminohydantoin (AHD), 3-amino-2-oxazolidinone (AOZ) and 3-amino-5-morpholinomethyl-2-oxazolinone (AMOZ) in gelatin Chinese medicine. A C18 column with the mobile phase consisting of acetonitrile and 5 mmol/L ammonium acetate in water was used to separate these nitrofuran metabolites. The limit of detection of SEM, AHD, AOZ and AMOZ were found to be 0.2 µg/kg, 0.3 µg/kg, 0.2 µg/kg and 0.2 µg/kg, whereas their limit of quantification were 0.6 µg/kg, 0.8 µg/kg, 0.6 µg/kg and 0.5 µg/kg. These nitrofuran metabolites exhibited a good linear standard curve (regression coefficients above 0.99) with a concentration range of 2 µg/L to 100 µg/L. Regarding extraction procedure, gelatin Chinese medicine was pre-treated with pepsin and then extracted using 5% formic acid (v/v) in acetonitrile. The resultant extract was purified through dispersive solid phase extraction using 1000 mg anhydrous sodium sulfate, 300 mg octadecyl carbon silica gel sorbent absorbent and 500 mg ethylenediamine-N-propyl carbon silica gel absorbent, and then further purified on Oasis PRiME HLB cartridges. The matrix effect was effectively eliminated after the clean-up procedure as confirmed by comparing the ratio of standard curves prepared by standards dissolved in both matrix solvent and 5 mmol/L ammonium acetate in water: acetonitrile (95:5, v/v). The recoveries of these nitrofuran metabolites under the 1 µg/kg, 2 µg/kg and 10 µg/kg spiking levels were between 77.4% and 95.6%. These metabolites after the extraction were stable at 4 °C for 24 h. The validated method was used to analyze the residue level of these nitrofuran metabolites in 25 gelatin Chinese medicines. Results showed that only one Colla Corii Asini sample contained SEM (2.52 µg/kg) and AOZ (6.27 µg/kg), whereas one Testudinis Carapacis et Plastri sample had SEM (1.27 µg/kg) and AMOZ (9.53 µg/kg).
Sigma-Aldrich
2-Nitrofuran, 97%
Supelco
AOZ, VETRANAL®, analytical standard