MilliporeSigma
  • FLIM-FRET-Based Structural Characterization of a Class-A GPCR Dimer in the Cell Membrane.
FLIM-FRET-Based Structural Characterization of a Class-A GPCR Dimer in the Cell Membrane.
Journal of molecular biology (2020-06-20)
Ju Yang, Zhou Gong, Yun-Bi Lu, Chan-Juan Xu, Tao-Feng Wei, Meng-Shi Yang, Tian-Wei Zhan, Yu-Hong Yang, Li Lin, Jianfeng Liu, Chun Tang, Wei-Ping Zhang
ABSTRACT
Class-A G protein-coupled receptors (GPCRs) are known to homo-dimerize in the membrane. Yet, methods to characterize the structure of GPCR dimer in the native environment are lacking. Accordingly, the molecular basis and functional relevance of the class-A GPCR dimerization remain unclear. Here, we present the dimeric structural model of GPR17 in the cell membrane. The dimer mainly involves transmembrane helix 5 (TM5) at the interface, with F229 in TM5, a critical residue. An F229A mutation makes GPR17 monomeric regardless of the expression level of the receptor. Monomeric mutants of GPR17 display impaired ERK1/2 activation and cannot be properly internalized upon agonist treatment. Conversely, the F229C mutant is cross-linked as a dimer and behaves like wild-type. Importantly, the GPR17 dimer structure has been modeled using sparse inter-protomer FRET distance restraints obtained from fluorescence lifetime imaging microscopy. The same approach can be applied to characterizing the interactions of other important membrane proteins in the cell.
Sigma-Aldrich
Uridine 5′-diphosphoglucose disodium salt hydrate from Saccharomyces cerevisiae, ≥98%
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%