MilliporeSigma
  • One-dimensional organic artificial multi-synapses enabling electronic textile neural network for wearable neuromorphic applications.
One-dimensional organic artificial multi-synapses enabling electronic textile neural network for wearable neuromorphic applications.
Science advances (2020-09-17)
Seonggil Ham, Minji Kang, Seonghoon Jang, Jingon Jang, Sanghyeon Choi, Tae-Wook Kim, Gunuk Wang
ABSTRACT
One-dimensional (1D) devices are becoming the most desirable format for wearable electronic technology because they can be easily woven into electronic (e-) textile(s) with versatile functional units while maintaining their inherent features under mechanical stress. In this study, we designed 1D fiber-shaped multi-synapses comprising ferroelectric organic transistors fabricated on a 100-μm Ag wire and used them as multisynaptic channels in an e-textile neural network for wearable neuromorphic applications. The device mimics diverse synaptic functions with excellent reliability even under 6000 repeated input stimuli and mechanical bending stress. Various NOR-type textile arrays are formed simply by cross-pointing 1D synapses with Ag wires, where each output from individual synapse can be integrated and propagated without undesired leakage. Notably, the 1D multi-synapses achieved up to ~90 and ~70% recognition accuracy for MNIST and electrocardiogram patterns, respectively, even in a single-layer neural network, and almost maintained regardless of the bending conditions.
Sigma-Aldrich
Pentacene, sublimed grade, ≥99.9% trace metals basis