MilliporeSigma
  • ASTILBIN: A PROMISING UNEXPLORED COMPOUND WITH MULTIDIMENSIONAL MEDICINAL AND HEALTH BENEFITS.
ASTILBIN: A PROMISING UNEXPLORED COMPOUND WITH MULTIDIMENSIONAL MEDICINAL AND HEALTH BENEFITS.
Pharmacological research (2020-05-15)
Abhishek Sharma, Sumeet Gupta, Samrat Chauhan, Anroop Nair, Prerna Sharma
ABSTRACT
Many flavonoids have various beneficial actions like anti-inflammatory, anti-carcinogenic properties and many other clinical conditions. Astilbin is one such flavanoid compound having many physiological as well as pharmacological actions. To summarize the important findings from the research conducted using astilbin having significance to its physiological and pharmacological activities as well as the patents filed using astilbin. Systematic review and compilation of the collected literature. An extensive investigation of literature was done using several worldwide electronic scientific databases like PUBMED, SCOPUS, Science Direct and Google Scholar etc. All the article available in the English language that used our compound of interest i.e. astilbin, on the basis of inclusion criteria decided were retrieved from these databases, thoroughly reviewed and were summarized. It has been established that astilbin can play a vital in the management of diseases associated with immune system. It also possesses antibacterial, anti-oxidative and hepatoprotective activity. These researches provide evidence that astilbin possesses great potential and thus can be utilized in the management of various disorders, thus establishing itself as a potential candidate for novel drug development. Also, there is still room for research on astilbin like it can be evaluated for anticancer potential, protective effect in various diabetic complications and many more. Overall observations from data suggested that astilbin is a promising compound and proved its efficacy in every preclinical study which is conducted till date. Some of the pharmacological activity is still unexplored. After successful preclinical trials, astilbin can go for further clinical trials.
Sigma-Aldrich
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)
Astilbin, phyproof® Reference Substance