MilliporeSigma
  • Hydrogen sulfide dilates the isolated retinal artery mainly via the activation of myosin phosphatase.
Hydrogen sulfide dilates the isolated retinal artery mainly via the activation of myosin phosphatase.
Life sciences (2020-05-27)
Ayça Toprak Semiz, Ayşegül Başak Teker, Kürşad Yapar, Birsel Sönmez Uydeş Doğan, Selçuk Takır
ABSTRACT
Hydrogen sulfide (H2S) is shown in ocular tissues and suggested to involve in the regulation of retinal circulation. However, the mechanism of H2S-induced relaxation on retinal artery is not clarified yet. Herein, we aimed to evaluate the role of several calcium (Ca2+) signaling and Ca2+ sensitization mechanisms in the relaxing effect of H2S donor, NaHS, on retinal arteries. Relaxing effects of NaHS (10-5-3 × 10-3M) were determined on precontracted retinal arteries in Ca2+ free medium as well as in the presence of the inhibitors of Ca2+ signaling and Ca2+ sensitization mechanisms. Additively, Ca2+ sensitivity of the contractile apparatus were evaluated by CaCl2-induced contractions in the presence of NaHS (3 × 10-3M). Functional experiments were furtherly assessed by protein and/or mRNA expressions, as appropriate. The relaxations to NaHS were preserved in Ca2+ free medium while NaHS pretreatment decreased the responsiveness to CaCl2. The inhibitors of plasmalemmal Ca2+-ATPase, sarcoplasmic-endoplasmic reticulum Ca2+-ATPase, Na+-Ca2+ ion-exchanger and myosin light chain kinase (MLCK) unchanged the relaxations to NaHS. Likewise, Ca2+ sensitization mechanisms including, rho kinase, protein kinase C and tyrosine kinase were unlikely to mediate the relaxation to NaHS in retinal artery. Whereas, a marked reduction was determined in NaHS-induced relaxations in the presence of MLCP inhibitor, calyculin A. Supportively, NaHS pretreatment significantly reduced phosphorylation of MYPT1-subunit of MLCP. The relaxing effect of NaHS in retinal artery is likely to be related to the activation of MLCP and partly, to decrement in Ca2+ sensitivity of contractile apparatus.