Merck
  • Evaluation of fetuin-A as a predictor of outcome after surgery for osteoporotic fracture of the proximal femur.
Evaluation of fetuin-A as a predictor of outcome after surgery for osteoporotic fracture of the proximal femur.
Archives of orthopaedic and trauma surgery (2019-12-21)
Claas Jasper Steffen, Philipp K E Herlyn, Norina Cornelius, Thomas Mittlmeier, Dagmar-C Fischer
ABSTRACT
In the elderly, osteoporotic fractures of the femur are associated with high morbidity and mortality. At the time of hospitalization and during pre-operative care identification of patients at risk for poor outcome despite an otherwise good clinical condition is challenging. We hypothesized that the serum concentration of fetuin-A during post-operative recovery might serve as a biomarker. During a 15 months period patients admitted to our hospital for treatment of a femur fracture were registered on a voluntary basis and irrespective of age, underlying diseases and therapy. For all patients enrolled in this registry, blood was sampled for additional laboratory analysis. Patients with osteoporotic fractures were invited for follow-up examination about 2 years later. At this time, the functional outcome and clinical performance together with the handgrip strength as a surrogate measure of overall strength were assessed and the occurrence of additional fractures was recorded. Results from these assessments and the absence or presence of subsequent fractures were combined as summary score of outcome (SSO) and overall physical performance (oPP), a score derived by principal component analysis and relying on normally distributed data, only. 39 of 96 eligible patients were deceased during the follow-up period, while 45 consented to participate in the study. Patients with a SSO ≥ 17 pts were younger and had higher fetuin-A serum concentrations (each p < 0.05) during hospitalization compared to those with a SSO below 8 pts. Age at follow-up together with total serum protein and fetuin-A levels during post-operative recovery had significant influence on SSO and oPP. Fetuin-A at time of follow-up examination was not associated with outcome. Our results are in favor of fetuin-A as an important factor for fracture healing but do not support an association between fetuin-A at time of follow-up examination and either SSO or oPP.
Sigma-Aldrich
Human AHSG / Alpha-2-HS-glycoprotein ELISA Kit, for serum, plasma and cell culture supernatants