MilliporeSigma
  • Integrating Two-Dimensional Gas and Liquid Chromatography-Mass Spectrometry for Untargeted Colorectal Cancer Metabolomics: A Proof-of-Principle Study.
Integrating Two-Dimensional Gas and Liquid Chromatography-Mass Spectrometry for Untargeted Colorectal Cancer Metabolomics: A Proof-of-Principle Study.
Metabolites (2020-08-29)
Fang Yuan, Seongho Kim, Xinmin Yin, Xiang Zhang, Ikuko Kato
ABSTRACT
Untargeted metabolomics is expected to lead to a better mechanistic understanding of diseases and thus applications of precision medicine and personalized intervention. To further increase metabolite coverage and achieve high accuracy of metabolite quantification, the present proof-of-principle study was to explore the applicability of integration of two-dimensional gas and liquid chromatography-mass spectrometry (GC × GC-MS and 2DLC-MS) platforms to characterizing circulating polar metabolome extracted from plasma collected from 29 individuals with colorectal cancer in comparison with 29 who remained cancer-free. After adjustment of multiple comparisons, 20 metabolites were found to be up-regulated and 8 metabolites were found to be down-regulated, which pointed to the dysregulation in energy metabolism and protein synthesis. While integrating the GC × GC-MS and 2DLC-MS data can dramatically increase the metabolite coverage, this study had a limitation in analyzing the non-polar metabolites. Given the small sample size, these results need to be validated with a larger sample size and with samples collected prior to diagnostic and treatment. Nevertheless, this proof-of-principle study demonstrates the potential applicability of integration of these advanced analytical platforms to improve discrimination between colorectal cancer cases and controls based on metabolite profiles in future studies.
Product Description
Supelco
C7 - C30 Saturated Alkanes, certified reference material, 1000 μg/mL each component in hexane