Merck
  • Correction: Nucleobases, nucleosides, and nucleotides: versatile biomolecules for generating functional nanomaterials.
Correction: Nucleobases, nucleosides, and nucleotides: versatile biomolecules for generating functional nanomaterials.
Chemical Society reviews (2018-01-18)
Fang Pu, Jinsong Ren, Xiaogang Qu
ABSTRACT
Correction for 'Nucleobases, nucleosides, and nucleotides: versatile biomolecules for generating functional nanomaterials' by Fang Pu et al., Chem. Soc. Rev., 2018, DOI: .
Sigma-Aldrich
5′-Nucleotides & Nucleosides