MilliporeSigma
Epinephrine responsiveness is reduced in livers from trained mice.
Physiological reports (2020-02-16)
Hana A Dibe, Logan K Townsend, Greg L McKie, David C Wright
ABSTRACT
The liver is the primary metabolic organ involved in the endogenous production of glucose through glycogenolysis and gluconeogenesis. Hepatic glucose production (HGP) is increased via neural-hormonal mechanisms such as increases in catecholamines. To date, the effects of prior exercise training on the hepatic response to epinephrine have not been fully elucidated. To examine the role of epinephrine signaling on indices of HGP in trained mice, male C57BL/6 mice were either subjected to 12 days of voluntary wheel running or remained sedentary. Epinephrine, or vehicle control, was injected intraperitoneally on day 12 prior to sacrifice with blood glucose being measured 15 min postinjection. Epinephrine caused a larger glucose response in sedentary mice and this was paralleled by a greater reduction in liver glycogen in sedentary compared to trained mice. There was a main effect of epinephrine to increase the phosphorylation of protein kinase-A (p-PKA) substrates in the liver, which was driven by increases in the sedentary, but not trained, mice. Similarly, epinephrine-induced increases in the mRNA expression of hepatic adrenergic receptors (Adra1/2a, Adrb1), and glucose-6-phosphatase (G6pc) were greater in sedentary compared to trained mice. The mRNA expression of cAMP-degrading enzymes phosphodiesterase 3B and 4B (Pde3b, Pde4b) was greater in trained compared to sedentary mice. Taken together, our data suggest that prior exercise training reduces the liver's response to epinephrine. This could be beneficial in the context of training-induced glycogen sparing during exercise.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)