MilliporeSigma
  • Homocysteine induces mesangial cell apoptosis via activation of p38-mitogen-activated protein kinase.
Homocysteine induces mesangial cell apoptosis via activation of p38-mitogen-activated protein kinase.
Kidney international (2006-12-07)
S Shastry, A J Ingram, J W Scholey, L R James
ABSTRACT
Hyperhomocysteinemia is prevalent among patients with chronic kidney disease (CKD) and has been linked to progressive kidney and vascular diseases. Increased glomerular mesangial cell (MC) turnover, including proliferation and apoptosis, is a hallmark of CKD. Activation of p38-mitogen-activated protein kinase (p38-MAPK) has been linked to apoptosis in many cell lines. Accordingly, we studied the effect of homocysteine (Hcy) on MC p38-MAPK signalling and apoptosis. Hcy (50 microM/24 h) increased MC apoptosis as determined by terminal deoxynucleotidyl transferase-mediated deoxyuridine triphosphate (dUTP) nick end labelling (TUNEL) and single-stranded DNA (ssDNA) analysis. In addition to increases in pro-caspase-3 protein and caspase-3 activity, cells exposed to Hcy manifested enhanced reactive oxygen species content. Hcy increased p38-MAPK activity (fivefold), with maximal effect at 50 microM and 20 min; p38-MAPK activation was attenuated by N-acetylcysteine (Nac) and catalase (Cat), further indicating that the effect was via oxidative stress. Confocal microscopy revealed activation and nuclear translocation of p38-MAPK that was attenuated by Cat. In addition, Hcy-induced apoptosis as determined by TUNEL and ssDNA assay was abrogated by Nac, Cat, and SB203580 (p38-MAPK inhibitor). We conclude that in MC, Hcy (i) activates p38-MAPK and increases p38MAPK nuclear translocation via an oxidative stress dependent mechanism and (ii) induces DNA damage and apoptosis that is dependent on oxidative stress and p38-MAPK activation.
Sigma-Aldrich
Caspase-3 Cellular Activity Assay Kit