MilliporeSigma
  • Circ_0000285 regulates proliferation, migration, invasion and apoptosis of osteosarcoma by miR-409-3p/IGFBP3 axis.
Circ_0000285 regulates proliferation, migration, invasion and apoptosis of osteosarcoma by miR-409-3p/IGFBP3 axis.
Cancer cell international (2020-10-13)
Zhisheng Long, Feipeng Gong, Yuxu Li, Zhiqiang Fan, Jingtang Li
ABSTRACT
Circular RNAs (circRNAs) are important regulators in the pathogenesis of diseases and affects the occurrence and development of diseases. However, the role of circRNAs in osteosarcoma (OS) has not been fully elucidated. The expression of circ_0000285, miR-409-3p and insulin-like growth factor binding protein 3 (IGFBP3) was detected using quantitative real-time PCR (qRT-PCR). The protein level of IGFBP3 was measured using western blot. CCK-8 and colony formation assays were used to determine cell proliferation. Flow cytometry was applied to measure cell cycle and cell apoptosis. Transwell assay was used to assess cell invasion and migration. Dual-luciferase reporter assay and RNA Binding Protein Immunoprecipitation (RIP) assay were performed to determine the relationship among circ_0000285, miR-409-3p and IGFBP3. The animal experiments were performed to determine the function of circ_0000285 in vivo. In this study, we found that the expression of circ_0000285 was significantly increased in OS tissues and cells and was enriched in the cytoplasm. Knockdown of circ_0000285 inhibited OS growth in vitro and in vivo. Moreover, miR-409-3p was a target miRNA of circ_0000285 and miR-409-3p targets to IGFBP3 in OS. Besides, circ_0000285 could promote proliferation, migration, invasion and inhibit apoptosis of osteosarcoma by miR-409-3p/IGFBP3 axis. In this study, circ_0000285 regulated proliferation, migration, invasion and apoptosis of OS cells by miR-409-3p/IGFBP3 axis, implying that circ_0000285 was a potential target for OS therapy.
Sigma-Aldrich
QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), fluorimetric, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with fluorescent detection.