MilliporeSigma
  • MH-76, a Novel Non-Quinazoline α1-Adrenoceptor Antagonist, but Not Prazosin Reduces Inflammation and Improves Insulin Signaling in Adipose Tissue of Fructose-Fed Rats.
MH-76, a Novel Non-Quinazoline α1-Adrenoceptor Antagonist, but Not Prazosin Reduces Inflammation and Improves Insulin Signaling in Adipose Tissue of Fructose-Fed Rats.
Pharmaceuticals (Basel, Switzerland) (2021-06-03)
Monika Kubacka, Szczepan Mogilski, Monika Zadrożna, Barbara Nowak, Małgorzata Szafarz, Bartosz Pomierny, Henryk Marona, Anna Waszkielewicz, Wojciech Jawień, Jacek Sapa, Marek Bednarski, Joanna Knutelska, Magdalena Kotańska
ABSTRACT
Quinazoline α1-adrenoceptors antagonists have been shown to exert moderately favorable effects on the metabolic profile in hypertensive patients. However, based on AntiHypertensive and Lipid-Lowering Treatment to Prevent Heart Attack Trial (ALLHAT) results, they are no longer recommended as a first line therapy of hypertension. Recent studies have shown that quinazoline-based α1-adrenoceptors antagonists (prazosin, doxazosin) induce the apoptosis and necrosis, which may be responsible for ALLHAT outcomes; however, these effects were proven to be independent of α1-adrenoceptor blockade and were associated with the presence of quinazoline moiety. MH-76 (1-[3-(2,6-dimethylphenoxy)propyl]-4-(2-methoxyphenyl)piperazine hydrochloride)) is a non-quinazoline α1-adrenoceptor antagonist which, in fructose-fed rats, exerted antihypertensive effect, and, contrary to prazosin, reduced insulin resistance and abdominal adiposity. In this study we aimed to further investigate and compare the effects of MH-76 and prazosin on inflammation in adipose tissue of fructose-fed rats. Abdominal adipose tissue was collected from four groups of fructose-fed rats (Control, Fructose, Fructose + MH-76 and Fructose + Prazosin) and subjected to biochemical, histopathological and immunohistochemical studies. Moreover, selected tissue distribution studies were performed. Treatment with MH-76 but not with prazosin improved endothelial integrity, reduced adipose tissue inflammation and infiltration by immune cells, resulting in lowering leptin, MCP-1, IL-6, TNF-α and PAI-1 levels. In adipose tissue from Fructose + MH-76 animals, a higher amount of eosinophils accompanied with higher IL-4 concentration was observed. Treatment with MH-76 but not with prazosin markedly reduced phosphorylation of IRS-1 at Ser307. MH-76 may improve insulin signaling in adipose tissue by reducing the pro-inflammatory cytokine production and inhibiting the inflammatory cells recruitment. In contrast, in adipose tissue from animals treated with prazosin, the inflammatory effect was clearly enhanced.
Product Description
Millipore
MILLIPLEX® Rat Adipocyte Panel - Metabolism Assay