MilliporeSigma
  • Highly conserved extracellular residues mediate interactions between pore-forming and regulatory subunits of the yeast Ca2+ channel related to the animal VGCC/NALCN family.
Highly conserved extracellular residues mediate interactions between pore-forming and regulatory subunits of the yeast Ca2+ channel related to the animal VGCC/NALCN family.
The Journal of biological chemistry (2020-07-22)
Takuto Hayashi, Keita Oishi, Midori Kimura, Kazuko Iida, Hidetoshi Iida
ABSTRACT
Yeasts and fungi generate Ca2+ signals in response to environmental stresses through Ca2+ channels essentially composed of Cch1 and Mid1. Cch1 is homologous to the pore-forming α1 subunit of animal voltage-gated Ca2+ channels (VGCCs) and sodium leak channels nonselective (NALCNs), whereas Mid1 is a membrane-associated protein similar to the regulatory α2/δ subunit of VGCCs and the regulatory subunit of NALCNs. Although the physiological roles of Cch1/Mid1 channels are known, their molecular regulation remains elusive, including subunit interactions regulating channel functionality. Herein, we identify amino acid residues involved in interactions between the pore-forming Cch1 subunit and the essential regulatory Mid1 subunit of Saccharomyces cerevisiae In vitro mutagenesis followed by functional assays and co-immunoprecipitation experiments reveal that three residues present in a specific extracellular loop in the repeat III region of Cch1 are required for interaction with Mid1, and that one essential Mid1 residue is required for interaction with Cch1. Importantly, these residues are necessary for Ca2+ channel activity and are highly conserved in fungal and animal counterparts. We discuss that this unique subunit interaction-based regulatory mechanism for Cch1 differs from that of VGCCs/NALCNs.