MilliporeSigma
  • A class of potent tyrosinase inhibitors: alkylidenethiosemicarbazide compounds.
A class of potent tyrosinase inhibitors: alkylidenethiosemicarbazide compounds.
European journal of medicinal chemistry (2008-06-06)
Jinbing Liu, Rihui Cao, Wei Yi, Chunming Ma, Yiqian Wan, Binhua Zhou, Lin Ma, Huacan Song
ABSTRACT
A series of alkylidenethiosemicarbazide compounds were synthesized and their inhibitory effects on the diphenolase activity of mushroom tyrosinase were evaluated. The results showed that most of the synthesized compounds exhibited significant inhibitory activities. Especially, compound 1f was found to be the most potent inhibitor with IC(50) value of 0.086 microM, suggesting that further development of such compounds may be of interest.
Sigma-Aldrich
Tropolone, 98%
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%