MilliporeSigma
  • MiR-25-3p targets PTEN to regulate the migration, invasion, and apoptosis of esophageal cancer cells via the PI3K/AKT pathway.
MiR-25-3p targets PTEN to regulate the migration, invasion, and apoptosis of esophageal cancer cells via the PI3K/AKT pathway.
Bioscience reports (2020-09-29)
Liang Zhang, Zhuang Tong, Zhe Sun, Guolian Zhu, Erdong Shen, Yanfeng Huang
ABSTRACT
Esophageal cancer (EC) is one of the most common malignant tumors of the digestive system. MiR-25-3p was proved to be a biomarker for the diagnosis and treatment of many cancers. MiR-25-3p was found to be high expressed in the blood of EC patients. The aim of the present study was to explore the effect of miR-25-3p and its target gene on EC. miR-25-3p expression in the blood of EC patients and EC cells was detected by RT-qPCR. The target of miR-25-3p was identified by bioinformatics and luciferase reporter assay. After transfection, cell viability, apoptosis, migration, and invasion were detected by MTT, flow cytometry, wound healing, and transwell assays, respectively. The expressions of PTEN, Bax, Bcl-2, cleaved Caspase-3, p-PI3K, PI3K, p-AKT, and AKT were detected by Western blot. MiR-25-3p was high expressed in the blood of EC patients and EC cells. MiR-25-3p targeted PTEN and inhibited the expression of PTEN. MiR-25-3p mimic increased the viability, migration, invasion and the expressions of Bcl-2, and inhibited the apoptosis and the expression of Bax and cleaved caspase-3 in EC cells. MiR-25-3p mimic also enhanced the expressions of p-PI3K and p-AKT and the ratios of p-PI3K/PI3K and p-AKT/AKT in EC cells. PTEN overexpression not only had an opposite effect of miR-25-3p mimic, but also reversed the effect of miR-25-3p mimic on EC cells. MiR-25-3p targeted PTEN to promote the migration and invasion, and inhibit apoptosis of EC cells via the PI3K/AKT pathway, which might provide a new therapeutic target for EC treatment.
Sigma-Aldrich
ChemiScreen Recombinant Human Glucagon Receptor Membrane Preparation, Human glucagon / GCG GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.