MilliporeSigma
  • Copper(I)-Catalyzed Nitrile-Addition/N-Arylation Ring-Closure Cascade: Synthesis of 5,11-Dihydro-6H-indolo[3,2-c]quinolin-6-ones as Potent Topoisomerase-I Inhibitors.
Copper(I)-Catalyzed Nitrile-Addition/N-Arylation Ring-Closure Cascade: Synthesis of 5,11-Dihydro-6H-indolo[3,2-c]quinolin-6-ones as Potent Topoisomerase-I Inhibitors.
Journal of medicinal chemistry (2021-01-26)
Wen-Yun Hsueh, Ying-Shuan E Lee, Min-Sian Huang, Chin-Hung Lai, Yu-Sheng Gao, Jo-Chu Lin, Yu-Fen Chen, Chih-Lin Chang, Shan-Yen Chou, Shyh-Fong Chen, Yann-Yu Lu, Lien-Hsiang Chang, Shu Fu Lin, Yu-Hsiang Lin, Pi-Chen Hsu, Win-Yin Wei, Ya-Chi Huang, Yi-Feng Kao, Li-Wei Teng, Hung-Huang Liu, Ying-Chou Chen, Ta-Tung Yuan, Ya-Wen Chan, Po-Hsun Huang, Yu-Ting Chao, Shin-Yi Huang, Bo-Han Jian, Hsin-Yi Huang, Sheng-Chuan Yang, Tzu-Hao Lo, Guan-Ru Huang, Shao-Yun Wang, Her-Sheng Lin, Shih-Hsien Chuang, Jiann-Jyh Huang
ABSTRACT
In this paper, we present a copper(I)-catalyzed nitrile-addition/N-arylation ring-closure cascade for the synthesis of 5,11-dihydro-6H-indolo[3,2-c]quinolin-6-ones from 2-(2-bromophenyl)-N-(2-cyanophenyl)acetamides. Using CuBr and t-BuONa in dimethylformamide (DMF) as the optimal reaction conditions, the cascade reaction gave the target products, in high yields, with a good substrate scope. Application of the cascade reaction was demonstrated on the concise total syntheses of alkaloid isocryptolepine. Further optimization of the products from the cascade reaction led to 3-chloro-5,12-bis[2-(dimethylamino)ethyl]-5,12-dihydro-6H-[1,3]dioxolo[4',5':5,6]indolo[3,2-c]quinolin-6-one (2k), which exhibited the characteristic DNA topoisomerase-I inhibitory mechanism of action with potent in vitro anticancer activity. Compound 2k actively inhibited ARC-111- and SN-38-resistant HCT-116 cells and showed in vivo activity in mice bearing human HCT-116 and SJCRH30 xenografts. The interaction of 2k with the Top-DNA cleavable complex was revealed by docking simulations to guide the future optimization of 5,11-dihydro-6H-indolo[3,2-c]quinolin-6-ones as topoisomerase-I inhibitors.
Sigma-Aldrich
2-Aminobenzylamine, 98%