MilliporeSigma
  • Synthesized tissue-equivalent dielectric phantoms using salt and polyvinylpyrrolidone solutions.
Synthesized tissue-equivalent dielectric phantoms using salt and polyvinylpyrrolidone solutions.
Magnetic resonance in medicine (2017-11-22)
Carlotta Ianniello, Jacco A de Zwart, Qi Duan, Cem M Deniz, Leeor Alon, Jae-Seung Lee, Riccardo Lattanzi, Ryan Brown
ABSTRACT
To explore the use of polyvinylpyrrolidone (PVP) for simulated materials with tissue-equivalent dielectric properties. PVP and salt were used to control, respectively, relative permittivity and electrical conductivity in a collection of 63 samples with a range of solute concentrations. Their dielectric properties were measured with a commercial probe and fitted to a 3D polynomial in order to establish an empirical recipe. The material's thermal properties and MR spectra were measured. The empirical polynomial recipe (available at https://www.amri.ninds.nih.gov/cgi-bin/phantomrecipe) provides the PVP and salt concentrations required for dielectric materials with permittivity and electrical conductivity values between approximately 45 and 78, and 0.1 to 2 siemens per meter, respectively, from 50 MHz to 4.5 GHz. The second- (solute concentrations) and seventh- (frequency) order polynomial recipe provided less than 2.5% relative error between the measured and target properties. PVP side peaks in the spectra were minor and unaffected by temperature changes. PVP-based phantoms are easy to prepare and nontoxic, and their semitransparency makes air bubbles easy to identify. The polymer can be used to create simulated material with a range of dielectric properties, negligible spectral side peaks, and long T2 relaxation time, which are favorable in many MR applications. Magn Reson Med 80:413-419, 2018. © 2017 International Society for Magnetic Resonance in Medicine.
Sigma-Aldrich
Select agar