MilliporeSigma
  • The usefulness of the combined high-dose dexamethasone suppression test and desmopressin stimulation test in establishing the source of ACTH secretion in ACTH-dependent Cushing's syndrome.
The usefulness of the combined high-dose dexamethasone suppression test and desmopressin stimulation test in establishing the source of ACTH secretion in ACTH-dependent Cushing's syndrome.
Endocrine journal (2021-04-02)
Jingtao Qiao, Jiaqi Li, Weiwei Zhang, Chun Wang, Jianwei Li, Shu Jiang, Huiwen Tan, Yaxi Chen, Hui Liu, Bowen Cai, Yerong Yu
ABSTRACT
Bilateral inferior petrosal sinus sampling (BIPSS) is the current gold standard test for differentially diagnosing ACTH-dependent Cushing's syndrome (CS). However, BIPSS is an invasive procedure, and its availability is limited. We retrospectively analysed the 24-hour urinary free cortisol (UFC) level during the high-dose dexamethasone suppression test (HDDST) and plasma ACTH/cortisol levels after the desmopressin stimulation test (DDAVP test) in subjects with confirmed Cushing's disease (CD) (n = 92) and ectopic ACTH-dependent CS (EAS) (n = 16), and evaluated the positive predictive value (PPV) of the two combined-tests in the aetiological diagnosis of ACTH-dependent CS. The percent changes in UFC levels after the HDDST and in ACTH/cortisol levels after DDAVP administration relative to the corresponding basal levels and the area under the receiver operating characteristic (ROC) curve (AUC) were analysed. UFC suppression below 62.7% suggested a pituitary origin with a sensitivity (SE) of 80% (95% CI: 70-88) and a specificity (SP) of 80% (95% CI: 52-96). A threshold increase in the ACTH level after DDAVP stimulation of 44.6% identified CD with an SE of 91% (95% CI: 83-97) and an SP of 75% (95% CI: 48-93). The combination of both tests yielded an SE of 95.5% and PPV of 98.4% for CD, and significantly improved the efficiency of the differential diagnosis between CD and EAS. These dual non-invasive endocrine tests may substantially reduce the need for BIPSS in the etiological investigation of ACTH-dependent CS.
Sigma-Aldrich
Anti-AVPR1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody