MilliporeSigma
  • Constitutive activation of S1P receptors at the trans-Golgi network is required for surface transport carrier formation.
Constitutive activation of S1P receptors at the trans-Golgi network is required for surface transport carrier formation.
iScience (2021-11-23)
Taro Okada, Susumu Nishida, Lifang Zhang, Nesma Nabil Ibrahim Mohamed, Tianyou Wang, Takeshi Ijuin, Taketoshi Kajimoto, Shun-Ichi Nakamura
ABSTRACT
The importance of the G-protein βγ subunits in the regulation of cargo transport from the trans-Golgi network (TGN) to the plasma membrane (PM) is well accepted; however, the molecular mechanism underlying the G-protein activation at the TGN remains unclear. We show here that sphingosine 1-phosphate (S1P) receptors at the PM were trafficked to the TGN in response to a surface transport cargo, temperature-sensitive vesicular stomatitis virus glycoprotein tagged with green fluorescent protein accumulation in the Golgi. The receptor internalization occurred in an S1P-independent manner but required phosphorylation by G-protein receptor kinase 2 and β-arrestin association before internalization. Continuously activated S1P receptors in a manner dependent on S1P at the TGN kept transmitting G-protein signals including the βγ subunits supply necessary for transport carrier formation at the TGN destined for the PM.
Sigma-Aldrich
Sphingosine Kinase Inhibitor, The Sphingosine Kinase Inhibitor, also referenced under CAS 1177741-83-1, controls the biological activity of Sphingosine Kinase. This small molecule/inhibitor is primarily used for Cell Signaling applications.