MilliporeSigma
Serosal mesothelium retains vasculogenic potential.
Developmental dynamics : an official publication of the American Association of Anatomists (2007-10-20)
Michiya Kawaguchi, David M Bader, Bettina Wilm
ABSTRACT
Mesothelia comprise the epithelial covering of coelomic organs and line the cavities in which they are housed. Mesothelia contribute to the vasculature of the heart and the intestinal tract by developmental processes of epithelial-mesenchymal transition (EMT), migration, and differentiation into endothelial cells, vascular smooth muscle cells, and pericytes. Here, we establish a novel in vitro system to analyze the differentiative potential of mesothelia. Using explants from serosal mesothelium (the mesothelial covering of the gut), we demonstrate that much of the developmental program observed in embryonic mesothelia is retained in the adult structure. Namely, processes of epithelial spreading, EMT, and differentiation into smooth muscle cells from these cells are observed. Interestingly, we were unable to stimulate endothelial cell differentiation using serum or various signaling factors. Taken together, these data reveal that differentiated serosal cells retain vasculogenic potential and provide a generalizable model for future studies on the developmental and differentiative capacity of the mesothelial cell type.
Product Description
Supelco
1,1-Dichloroethylene, analytical standard