MilliporeSigma
Hypothesis--haptoglobin genotype and diabetic nephropathy.
Nature clinical practice. Nephrology (2007-05-26)
Farid M Nakhoul, Rachel Miller-Lotan, Huda Awaad, Rabea Asleh, Andrew P Levy
ABSTRACT
Vascular complications cause serious morbidity in patients with diabetes mellitus. Three such complications are nephropathy, retinopathy and accelerated atherosclerotic cardiovascular disease. There is currently scant evidence of a genetic marker that predicts which patients will have vascular complications. Oxidative stress has an important role in the development of diabetic vascular complications. Haptoglobin (Hp) is a hemoglobin-binding protein that has a major role in protecting against heme-driven oxidative stress. There are two common alleles for Hp (1 and 2) and, therefore, three common Hp genotypes: Hp 1-1, Hp 2-1, and Hp 2-2. The antioxidant protection provided by Hp is genotype-dependent; the protein encoded by Hp 1-1 provides superior antioxidant protection compared with that encoded by Hp 2-2. We have shown that diabetic individuals with Hp 2-2 are more likely to develop nephropathy, retinopathy, and cardiovascular disease than those with the Hp 2-1 or Hp 1-1 genotypes.