MilliporeSigma
  • Deletion of the protein tyrosine phosphatase gene PTPN2 in T-cell acute lymphoblastic leukemia.
Deletion of the protein tyrosine phosphatase gene PTPN2 in T-cell acute lymphoblastic leukemia.
Nature genetics (2010-05-18)
Maria Kleppe, Idoya Lahortiga, Tiama El Chaar, Kim De Keersmaecker, Nicole Mentens, Carlos Graux, Katrien Van Roosbroeck, Adolfo A Ferrando, Anton W Langerak, Jules P P Meijerink, François Sigaux, Torsten Haferlach, Iwona Wlodarska, Peter Vandenberghe, Jean Soulier, Jan Cools
ABSTRACT
PTPN2 (protein tyrosine phosphatase non-receptor type 2, also known as TC-PTP) is a cytosolic tyrosine phosphatase that functions as a negative regulator of a variety of tyrosine kinases and other signaling proteins. In agreement with its role in the regulation of the immune system, PTPN2 was identified as a susceptibility locus for autoimmune diseases. In this work, we describe the identification of focal deletions of PTPN2 in human T-cell acute lymphoblastic leukemia (T-ALL). Deletion of PTPN2 was specifically found in T-ALLs with aberrant expression of the TLX1 transcription factor oncogene, including four cases also expressing the NUP214-ABL1 tyrosine kinase. Knockdown of PTPN2 increased the proliferation and cytokine sensitivity of T-ALL cells. In addition, PTPN2 was identified as a negative regulator of NUP214-ABL1 kinase activity. Our study provides genetic and functional evidence for a tumor suppressor role of PTPN2 and suggests that expression of PTPN2 may modulate response to treatment.