MilliporeSigma
  • The inhibitory receptor PD-1 regulates IgA selection and bacterial composition in the gut.
The inhibitory receptor PD-1 regulates IgA selection and bacterial composition in the gut.
Science (New York, N.Y.) (2012-04-28)
Shimpei Kawamoto, Thinh H Tran, Mikako Maruya, Keiichiro Suzuki, Yasuko Doi, Yumi Tsutsui, Lucia M Kato, Sidonia Fagarasan
ABSTRACT
Immunoglobulin A (IgA) is essential to maintain the symbiotic balance between gut bacterial communities and the host immune system. Here we provide evidence that the inhibitory co-receptor programmed cell death-1 (PD-1) regulates the gut microbiota through appropriate selection of IgA plasma cell repertoires. PD-1 deficiency generates an excess number of T follicular helper (T(FH)) cells with altered phenotypes, which results in dysregulated selection of IgA precursor cells in the germinal center of Peyer's patches. Consequently, the IgAs produced in PD-1-deficient mice have reduced bacteria-binding capacity, which causes alterations of microbial communities in the gut. Thus, PD-1 plays a critical role in regulation of antibody diversification required for the maintenance of intact mucosal barrier.