MilliporeSigma
  • Resonance Raman study on complexes of medium-chain acyl-CoA dehydrogenase.
Resonance Raman study on complexes of medium-chain acyl-CoA dehydrogenase.
Journal of biochemistry (1992-06-01)
Y Nishina, K Sato, K Shiga, S Fujii, K Kuroda, R Miura
ABSTRACT
Resonance Raman (RR) spectra of the complex of pig kidney medium-chain acyl-CoA dehydrogenase with acetoacetyl-CoA and of the purple complex formed upon the addition of octanoyl-CoA to the dehydrogenase were obtained. RR spectra were also measured for the complexes prepared by using isotopically labeled compounds, i.e., [3-13C]-, [1,3-13C]-, and [2,4-13C2]acetoacetyl-CoA; [1-13C]octanoyl-CoA; the dehydrogenase reconstituted with [4a-13C]- and [4,10a-13C2]FAD. Both bands of oxidized flavin and acetoacetyl-CoA were resonance-enhanced in the 632.8 nm excited spectra of the acetoacetyl-CoA complex; this confirms that the broad long-wavelength absorption band is a charge-transfer absorption band between oxidized flavin and acetoacetyl-CoA. The 1,622 cm-1 band was assigned to the C(3)=O stretching mode coupling with the C(2)-H bending mode of the enolate form of acetoacetyl-CoA and the bands at 1,483 and 1,119 cm-1 were assigned to bands associated with the C(2)=C(1)-O- moiety. Both bands of fully reduced flavin and the substrate were resonance-enhanced in the 632.8 nm excited spectra of the purple complex. As the enzyme is already reduced, the substrate must be oxidized to octenoyl-CoA; the complex is a charge-transfer complex between the reduced enzyme and octenoyl-CoA. The low frequency value of the 1,577 cm-1 band, which is associated with the C(2)-C(1)=O moiety of the octenoyl-CoA, suggests that the enzyme-bound octenoyl-CoA has an appreciable contribution of C(2)=C(1)-O-.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Octanoic acid-1-13C, 99 atom % 13C