MilliporeSigma
  • Intestinal epithelial tight junction barrier regulation by autophagy-related protein ATG6/beclin 1.
Intestinal epithelial tight junction barrier regulation by autophagy-related protein ATG6/beclin 1.
American journal of physiology. Cell physiology (2019-03-21)
Morgan Wong, Ashwinkumar Subramenium Ganapathy, Eric Suchanec, Laura Laidler, Thomas Ma, Prashant Nighot
ABSTRACT
A defective tight junction (TJ) barrier is a key pathogenic factor for inflammatory bowel disease. Previously, we have shown that autophagy, a cell survival mechanism, enhances intestinal epithelial TJ barrier function. Autophagy-related protein-6 (ATG6/beclin 1), a key protein in the autophagy pathway, also plays a role in the endocytic pathway. The constitutive role of beclin 1 in the intestinal TJ barrier is not known. In Caco-2 cells, beclin 1 was found to be coimmunoprecipitated with the TJ protein occludin and colocalized with occludin on the membrane. Treatment of Caco-2 cells with beclin 1 peptide [transactivating regulatory protein (Tat)-beclin 1] reduced TJ barrier function. Activation of beclin 1 increased occludin endocytosis and reduced total occludin protein level. In contrast, beclin 1 siRNA transfection enhanced Caco-2 TJ barrier function. In pharmacologic and genetic autophagy inhibition studies, the constitutive function of beclin 1 in the TJ barrier was found to be autophagy independent. However, de novo induction of autophagy with starvation or rapamycin prevented Tat-beclin 1-induced increase in TJ permeability and reduction in occludin level. Induction of autophagy also resulted in reduced beclin 1-occludin association. In mouse colon, beclin 1 colocalized with occludin on the epithelial membrane. Perfusion of mouse colon with beclin 1 peptide caused an increase in colonic TJ permeability that was prevented by in vivo induction of autophagy. These findings show that beclin 1 plays a constitutive, autophagy-independent role in the regulation of intestinal TJ barrier function via endocytosis of occludin. Autophagy terminates constitutive beclin 1 function in the TJ barrier and enhances the TJ barrier.
Sigma-Aldrich
Earle′s Balanced Salts, With sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture