MilliporeSigma
  • Recovery of respiratory function in mdx mice co-treated with neutralizing interleukin-6 receptor antibodies and urocortin-2.
Recovery of respiratory function in mdx mice co-treated with neutralizing interleukin-6 receptor antibodies and urocortin-2.
The Journal of physiology (2018-08-31)
David P Burns, Leonie Canavan, Jane Rowland, Robin O'Flaherty, Molly Brannock, Sarah E Drummond, Dervla O'Malley, Deirdre Edge, Ken D O'Halloran
ABSTRACT
Impaired ventilatory capacity and diaphragm muscle weakness are prominent features of Duchenne muscular dystrophy, with strong evidence of attendant systemic and muscle inflammation. We performed a 2-week intervention in young wild-type and mdx mice, consisting of either injection of saline or co-administration of a neutralizing interleukin-6 receptor antibody (xIL-6R) and urocortin-2 (Ucn2), a corticotrophin releasing factor receptor 2 agonist. We examined breathing and diaphragm muscle form and function. Breathing and diaphragm muscle functional deficits are improved following xIL-6R and Ucn2 co-treatment in mdx mice. The functional improvements were associated with a preservation of mdx diaphragm muscle myosin heavy chain IIx fibre complement. The concentration of the pro-inflammatory cytokine interleukin-1β was reduced and the concentration of the anti-inflammatory cytokine interleukin-10 was increased in mdx diaphragm following drug co-treatment. Our novel findings may have implications for the development of pharmacotherapies for the dystrophinopathies with relevance for respiratory muscle performance and breathing. The mdx mouse model of Duchenne muscular dystrophy shows evidence of hypoventilation and pronounced diaphragm dysfunction. Six-week-old male mdx (n = 32) and wild-type (WT; n = 32) mice received either saline (0.9% w/v) or a co-administration of neutralizing interleukin-6 receptor antibodies (xIL-6R; 0.2 mg kg-1 ) and corticotrophin-releasing factor receptor 2 agonist (urocortin-2; 30 μg kg-1 ) subcutaneously over 2 weeks. Breathing and diaphragm muscle contractile function (ex vivo) were examined. Diaphragm structure was assessed using histology and immunofluorescence. Muscle cytokine concentration was determined using a multiplex assay. Minute ventilation and diaphragm muscle peak force at 100 Hz were significantly depressed in mdx compared with WT. Drug treatment completely restored ventilation in mdx mice during normoxia and significantly increased mdx diaphragm force- and power-generating capacity. The number of centrally nucleated muscle fibres and the areal density of infiltrates and collagen content were significantly increased in mdx diaphragm; all indices were unaffected by drug co-treatment. The abundance of myosin heavy chain (MyHC) type IIx fibres was significantly decreased in mdx diaphragm; drug co-treatment preserved MyHC type IIx complement in mdx muscle. Drug co-treatment increased the cross-sectional area of MyHC type I and IIx fibres in mdx diaphragm. The cytokines IL-1β, IL-6, KC/GRO and TNF-α were significantly increased in mdx diaphragm compared with WT. Drug co-treatment significantly decreased IL-1β and increased IL-10 in mdx diaphragm. Drug co-treatment had no significant effect on WT diaphragm muscle structure, cytokine concentrations or function. Recovery of breathing and diaphragm force in mdx mice was impressive in our studies, with implication for human dystrophinopathies.
Sigma-Aldrich
Urocortin II mouse