MilliporeSigma
  • Sustained Alzheimer's amyloid pathology in myeloid differentiation protein-88-deficient APPswe/PS1 mice.
Sustained Alzheimer's amyloid pathology in myeloid differentiation protein-88-deficient APPswe/PS1 mice.
Neuro-degenerative diseases (2013-11-07)
Y Goll, U Bekenstein, S Barbash, D S Greenberg, R Zangen, S Shoham, H Soreq
ABSTRACT
Most Alzheimer's disease (AD) cases arise sporadically and may involve innate immune activation of microglial expressed Toll-like receptors regulated through the myeloid differentiation protein 88 (MyD88) pathway. It was the aim of this study to test the innate immune involvement in AD pathology. We mated APPsw/PS1ΔE9 mice with MyD88-deficient mice. Progeny mice had similar levels of soluble amyloid-β peptides, amyloid plaque density and neuroimmune staining patterns. However, double-transgenic mice did show a significantly reduced life expectancy. Our findings indicate that impaired innate immune responses may play a role in AD pathology.
Product Description
Millipore
Human Amyloid β42 Brain ELISA, This Human Amyloid β42 Brain ELISA is used to measure & quantify Amyloid β42 levels in Neuroscience research.