MilliporeSigma
Buccal micronucleus cytome assay.
Nature protocols (2009-05-16)
Philip Thomas, Nina Holland, Claudia Bolognesi, Micheline Kirsch-Volders, Stefano Bonassi, Errol Zeiger, Siegfried Knasmueller, Michael Fenech
ABSTRACT
The Buccal Micronucleus Cytome (BMCyt) assay is a minimally invasive method for studying DNA damage, chromosomal instability, cell death and the regenerative potential of human buccal mucosal tissue. This method is increasingly used in molecular epidemiological studies for investigating the impact of nutrition, lifestyle factors, genotoxin exposure and genotype on DNA damage, chromosome malsegregation and cell death. The biomarkers measured in this assay have been associated with increased risk of accelerated ageing, cancer and neurodegenerative diseases. This protocol describes one of the current established methods for buccal cell collection using a small-headed toothbrush, the generation of a single-cell suspension, slide preparation using cytocentrifugation, fixation and staining using Feulgen and Light Green for both bright field and fluorescence microscopic analysis. The scoring criteria for micronuclei and other nuclear anomalies are also described in detail. The protocol in its current form takes approximately 4 h to complete from the time of buccal cell collection to the generation of stained slides for microscopic analysis.
Sigma-Aldrich
Ethylenediaminetetraacetic acid tetrasodium salt hydrate, ≥99.0%