MilliporeSigma
  • 4-Methylthio-3-butenyl isothiocyanate (MTBITC) induced apoptotic cell death and G2/M cell cycle arrest via ROS production in human esophageal epithelial cancer cells.
4-Methylthio-3-butenyl isothiocyanate (MTBITC) induced apoptotic cell death and G2/M cell cycle arrest via ROS production in human esophageal epithelial cancer cells.
The Journal of toxicological sciences (2019-02-07)
Tadashi Hirata, Young-Man Cho, Isamu Suzuki, Takeshi Toyoda, Jun-Ichi Akagi, Yasushi Nakamura, Satoshi Numazawa, Kumiko Ogawa
ABSTRACT
To investigate the chemopreventive mechanisms of 4-Methylthio-3-butenyl isothiocyanate (MTBITC), we analyzed cell viability, cell cycle distribution, and expression levels for cell cycle and apoptosis-related proteins in MTBITC-treated malignant esophageal KYSE510 cells, with and without the reactive oxygen species (ROS) scavenger N-acethyl-L-Cysteine (NAC). MTBITC dose-dependently reduced cell viability and Bcl2 protein expression, while it induced cleavages of caspase-3, caspase-9, and PARP-1, suggesting that reduced cell viability occurred through the mitochondrial apoptotic pathway in KYSE510 cells. In cell cycle distribution analysis, MTBITC (20-40 µM) induced cell cycle arrest at G2/M phase. Furthermore, MTBITC induced Chk1 and Akt phosphorylations and decreased p27 protein expression. Both apoptotic- and cell cycle-related changes induced by MTBITC treatment were abolished by NAC. These results suggest that MTBITC has chemopreventive potential for esophageal carcinogenesis by elimination of cancer cells via induction of mitochondrial apoptotic cell death, G2/M cell cycle arrest, and ROS production.
Sigma-Aldrich
Erucin, ≥98% (HPLC)