MilliporeSigma
  • Overexpressed DDX3x promotes abdominal aortic aneurysm formation and activates AKT in ApoE knockout mice.
Overexpressed DDX3x promotes abdominal aortic aneurysm formation and activates AKT in ApoE knockout mice.
Biochemical and biophysical research communications (2022-10-16)
Yifei Zhou, Hao Chai, Yuntao Hu, Renjie Liu, Hongwei Jiang, Rui Fan, Wen Chen, Xin Chen, Fuhua Huang
ABSTRACT
In recent years, abdominal aortic aneurysm (AAA) lesions have become one of the important diseases that threaten public health. Related studies have confirmed that the occurrence of abdominal aortic aneurysms is related to inflammatory stress, cell apoptosis, and elastic fiber degradation. DDX3x is thought to interact with inflammasomes such as NLRP3 to aggravate the process of the inflammatory response, but its role in the occurrence of AAA remains unclear. Since DDX3x is indispensable in animal embryonic growth, we used an adeno-associated virus to construct gene-overexpressing mice to induce aneurysm development through AngII infusion. The results indicated that the incidence of aneurysms, inflammatory cell infiltration, vascular smooth muscle cell transformation, and oxidative stress levels were significantly increased under the condition of DDX3x overexpression. At the signaling level, activation of the AKT pathway exacerbates aneurysm formation. Taken together, we believe that DDX3x plays a key role in the development of aneurysms and may be a new target for the treatment of aneurysm progression.
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder