MilliporeSigma
A structural basis for prion strain diversity.
Nature chemical biology (2023-01-17)
Szymon W Manka, Adam Wenborn, Jemma Betts, Susan Joiner, Helen R Saibil, John Collinge, Jonathan D F Wadsworth
ABSTRACT
Recent cryogenic electron microscopy (cryo-EM) studies of infectious, ex vivo, prion fibrils from hamster 263K and mouse RML prion strains revealed a similar, parallel in-register intermolecular β-sheet (PIRIBS) amyloid architecture. Rungs of the fibrils are composed of individual prion protein (PrP) monomers that fold to create distinct N-terminal and C-terminal lobes. However, disparity in the hamster/mouse PrP sequence precludes understanding of how divergent prion strains emerge from an identical PrP substrate. In this study, we determined the near-atomic resolution cryo-EM structure of infectious, ex vivo mouse prion fibrils from the ME7 prion strain and compared this with the RML fibril structure. This structural comparison of two biologically distinct mouse-adapted prion strains suggests defined folding subdomains of PrP rungs and the way in which they are interrelated, providing a structural definition of intra-species prion strain-specific conformations.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution