MilliporeSigma
  • Identification of human zonulin, a physiological modulator of tight junctions, as prehaptoglobin-2.
Identification of human zonulin, a physiological modulator of tight junctions, as prehaptoglobin-2.
Proceedings of the National Academy of Sciences of the United States of America (2009-10-07)
Amit Tripathi, Karen M Lammers, Simeon Goldblum, Terez Shea-Donohue, Sarah Netzel-Arnett, Marguerite S Buzza, Toni M Antalis, Stefanie N Vogel, Aiping Zhao, Shiqi Yang, Marie-Claire Arrietta, Jon B Meddings, Alessio Fasano
ABSTRACT
Increased intestinal permeability (IP) has emerged recently as a common underlying mechanism in the pathogenesis of allergic, inflammatory, and autoimmune diseases. The characterization of zonulin, the only physiological mediator known to regulate IP reversibly, has remained elusive. Through proteomic analysis of human sera, we have now identified human zonulin as the precursor for haptoglobin-2 (pre-HP2). Although mature HP is known to scavenge free hemoglobin (Hb) to inhibit its oxidative activity, no function has ever been ascribed to its uncleaved precursor form. We found that the single-chain zonulin contains an EGF-like motif that leads to transactivation of EGF receptor (EGFR) via proteinase-activated receptor 2 (PAR(2)) activation. Activation of these 2 receptors was coupled to increased IP. The siRNA-induced silencing of PAR(2) or the use of PAR(2)(-/-) mice prevented loss of barrier integrity. Proteolytic cleavage of zonulin into its alpha(2)- and beta-subunits neutralized its ability to both activate EGFR and increase IP. Quantitative gene expression revealed that zonulin is overexpressed in the intestinal mucosa of subjects with celiac disease. To our knowledge, this is the initial example of a molecule that exerts a biological activity in its precursor form that is distinct from the function of its mature form. Our results therefore characterize zonulin as a previously undescribed ligand that engages a key signalosome involved in the pathogenesis of human immune-mediated diseases that can be targeted for therapeutic interventions.
Sigma-Aldrich
Trypsin Agarose, buffered aqueous suspension, from bovine pancreas trypsin