MilliporeSigma
  • Secretion of mitochondrial DNA via exosomes promotes inflammation in Behçet's syndrome.
Secretion of mitochondrial DNA via exosomes promotes inflammation in Behçet's syndrome.
The EMBO journal (2023-09-04)
Hachiro Konaka, Yasuhiro Kato, Toru Hirano, Kohei Tsujimoto, JeongHoon Park, Taro Koba, Wataru Aoki, Yusei Matsuzaki, Masayasu Taki, Shohei Koyama, Eri Itotagawa, Tatsunori Jo, Takehiro Hirayama, Taro Kawai, Ken J Ishii, Mitsuyoshi Ueda, Shigehiro Yamaguchi, Shizuo Akira, Takayoshi Morita, Yuichi Maeda, Masayuki Nishide, Sumiyuki Nishida, Yoshihito Shima, Masashi Narazaki, Hyota Takamatsu, Atsushi Kumanogoh
ABSTRACT
Mitochondrial DNA (mtDNA) leakage into the cytoplasm can occur when cells are exposed to noxious stimuli. Specific sensors recognize cytoplasmic mtDNA to promote cytokine production. Cytoplasmic mtDNA can also be secreted extracellularly, leading to sterile inflammation. However, the mode of secretion of mtDNA out of cells upon noxious stimuli and its relevance to human disease remain unclear. Here, we show that pyroptotic cells secrete mtDNA encapsulated within exosomes. Activation of caspase-1 leads to mtDNA leakage from the mitochondria into the cytoplasm via gasdermin-D. Caspase-1 also induces intraluminal membrane vesicle formation, allowing for cellular mtDNA to be taken up and secreted as exosomes. Encapsulation of mtDNA within exosomes promotes a strong inflammatory response that is ameliorated upon exosome biosynthesis inhibition in vivo. We further show that monocytes derived from patients with Behçet's syndrome (BS), a chronic systemic inflammatory disorder, show enhanced caspase-1 activation, leading to exosome-mediated mtDNA secretion and similar inflammation pathology as seen in BS patients. Collectively, our findings support that mtDNA-containing exosomes promote inflammation, providing new insights into the propagation and exacerbation of inflammation in human inflammatory diseases.
Sigma-Aldrich
Nigericin sodium salt Ready Made Solution, 5 mg/mL (DMSO:ethanol 1:1)