MilliporeSigma
  • Auxin regulates bulbil initiation by mediating sucrose metabolism in Lilium lancifolium.
Auxin regulates bulbil initiation by mediating sucrose metabolism in Lilium lancifolium.
Horticulture research (2024-05-06)
Yin Xin, Xi Chen, Jiahui Liang, Shaokun Wang, Wenqiang Pan, Jingxiang Wu, Mingfang Zhang, Michele Zaccai, Xiaonan Yu, Xiuhai Zhang, Jian Wu, Yunpeng Du
ABSTRACT
Lily bulbils, which serve as advantageous axillary organs for vegetative propagation, have not been extensively studied in terms of the mechanism of bulbil initiation. The functions of auxin and sucrose metabolism have been implicated in axillary organ development, but their relationship in regulating bulbil initiation remains unclear. In this study, exogenous indole-3-acetic acid (IAA) treatment increased the endogenous auxin levels at leaf axils and significantly decreased bulbil number, whereas treatment with the auxin polar transport inhibitor N-1-naphthylphthalamic acid (NPA), which resulted in a low auxin concentration at leaf axils, stimulated bulbil initiation and increased bulbil number. A low level of auxin caused by NPA spraying or silencing of auxin biosynthesis genes YUCCA FLAVIN MONOOXYGENASE-LIKE 6 (LlYUC6) and TRYPTOPHAN AMINOTRANSFERASERELATED 1 (LlTAR1) facilitated sucrose metabolism by activating the expression of SUCROSE SYNTHASES 1 (LlSusy1) and CELL WALL INVERTASE 2 (LlCWIN2), resulting in enhanced bulbil initiation. Silencing LlSusy1 or LlCWIN2 hindered bulbil initiation. Moreover, the transcription factor BASIC HELIX-LOOP-HELIX 35 (LlbHLH35) directly bound the promoter of LlSusy1, but not the promoter of LlCWIN2, and activated its transcription in response to the auxin content, bridging the gap between auxin and sucrose metabolism. In conclusion, our results reveal that an LlbHLH35-LlSusy1 module mediates auxin-regulated sucrose metabolism during bulbil initiation.
Sigma-Aldrich
Anti-Auxin antibody, Mouse monoclonal, clone 1E11-C11, purified from hybridoma cell culture