MilliporeSigma
  • Vitamin D Receptor/Vitamin D Response Element Directly Modulate Nestin Transcription to Ameliorate PAN-Induced Podocyte Morphological Changes.
Vitamin D Receptor/Vitamin D Response Element Directly Modulate Nestin Transcription to Ameliorate PAN-Induced Podocyte Morphological Changes.
Nephron (2022-05-09)
Qian Zhang, Xinxin Jiang, Donghai Wen, Yulin Zhang, Jianping Mao, Li Ni, Jing Chen
ABSTRACT
The change of podocyte morphology is a pathologic feature of chronic kidney disease. Several studies have suggested that vitamin D plays a role in the protection of podocytes, but the underlying mechanism remains unclear. The effects of paricalcitol on podocyte injury were tested in a puromycin aminonucleoside (PAN)-induced rat model and cultured mouse podocytes. Proteinuria, podocyte foot process (FP) effacement, and the expression of nestin and vitamin D receptor (VDR) were evaluated. VDR-siRNA or plasmids containing VDR-shRNA were transfected into podocytes to silence VDR expression. Chromatin immunoprecipitation (ChIP) and luciferase reporter assays were performed to verify the connection between VDR and nestin gene expression. Paricalcitol significantly alleviated proteinuria and podocyte FP effacement in PAN-induced nephrosis, which was accompanied by increased VDR expression in the glomeruli. Paricalcitol also inhibited PAN-induced nestin overexpression in the glomeruli. In an in vivo study, PAN significantly inhibited VDR protein expression, stimulated nestin protein expression, and resulted in nestin filament derangement in mouse podocytes, while paricalcitol treatment abolished these effects. In contrast, downregulation of VDR resulted in derangement and overexpression of nestin. ChIP assays demonstrated the presence of a vitamin D response element (VDRE) in the nestin promoter, and paricalcitol enhanced the binding of VDR to VDRE. Furthermore, luciferase reporter assays of the nestin promoter fragment showed that paricalcitol effectively repressed nestin reporter gene expression after PAN treatment, and mutation of VDRE abolished this effect. Paricalcitol directly regulates nestin transcription through the interaction of VDR/VDRE, thereby preventing morphological changes of podocytes in PAN nephropathy.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody