MilliporeSigma
  • The effects of growth hormone (GH) treatment on GH and insulin/IGF-1 signaling in long-lived Ames dwarf mice.
The effects of growth hormone (GH) treatment on GH and insulin/IGF-1 signaling in long-lived Ames dwarf mice.
The journals of gerontology. Series A, Biological sciences and medical sciences (2009-11-13)
Michal M Masternak, Jacob A Panici, Feiya Wang, Zhihui Wang, Adam Spong
ABSTRACT
The disruption of the growth hormone (GH) axis in mice promotes insulin sensitivity and is strongly correlated with extended longevity. Ames dwarf (Prop1(df), df/df) mice are GH, prolactin (PRL), and thyrotropin (TSH) deficient and live approximately 50% longer than their normal siblings. To investigate the effects of GH on insulin and GH signaling pathways, we subjected these dwarf mice to twice-daily GH injections (6 microg/g/d) starting at the age of 2 weeks and continuing for 6 weeks. This produced the expected activation of the GH signaling pathway and stimulated somatic growth of the Ames dwarf mice. However, concomitantly with increased growth and increased production of insulinlike growth factor-1, the GH treatment strongly inhibited the insulin signaling pathway by decreasing insulin sensitivity of the dwarf mice. This suggests that improving growth of these animals may negatively affect both their healthspan and longevity by causing insulin resistance.
Sigma-Aldrich
Insulin from porcine pancreas, powder, ≥27 USP units/mg