MilliporeSigma
  • Paradoxical imbalance between activated lymphocyte protein synthesis capacity and rapid division rate.
Paradoxical imbalance between activated lymphocyte protein synthesis capacity and rapid division rate.
eLife (2024-03-21)
Mina O Seedhom, Devin Dersh, Jaroslav Holly, Mariana Pavon-Eternod, Jiajie Wei, Matthew Angel, Lucas Shores, Alexandre David, Jefferson Santos, Heather Hickman, Jonathan W Yewdell
ABSTRACT
Rapid lymphocyte cell division places enormous demands on the protein synthesis machinery. Flow cytometric measurement of puromycylated ribosome-associated nascent chains after treating cells or mice with translation initiation inhibitors reveals that ribosomes in resting lymphocytes in vitro and in vivo elongate at typical rates for mammalian cells. Intriguingly, elongation rates can be increased up to 30% by activation in vivo or fever temperature in vitro. Resting and activated lymphocytes possess abundant monosome populations, most of which actively translate in vivo, while in vitro, nearly all can be stalled prior to activation. Quantitating lymphocyte protein mass and ribosome count reveals a paradoxically high ratio of cellular protein to ribosomes insufficient to support their rapid in vivo division, suggesting that the activated lymphocyte proteome in vivo may be generated in an unusual manner. Our findings demonstrate the importance of a global understanding of protein synthesis in lymphocytes and other rapidly dividing immune cells.
Product Description
Millipore
Emetine, Dihydrochloride, Principal alkaloid of ipecac, isolated from the ground roots of Uragoga ipecacuanha.