MilliporeSigma
  • Design and synthesis of a beta-lactamase activated 5-fluorouracil prodrug.
Design and synthesis of a beta-lactamase activated 5-fluorouracil prodrug.
Bioorganic & medicinal chemistry letters (2009-01-27)
Ryan M Phelan, Marc Ostermeier, Craig A Townsend
ABSTRACT
An efficient synthesis of a 5-fluorouracil-cephalosporin prodrug is described for use against colorectal and other cancers in antibody and gene-directed therapies. The compound shows stability in aqueous media until specifically activated by beta-lactamase (betaL). The kinetic parameters of the 5-fluorouracil-cephalosporin conjugate were determined in the presence of Enterobacter cloacae P99 betaL (ECl betaL) revealing a K(m)=95.4 microM and V(max)=3.21 microMol min(-1) mg(-1). The data compare favorably to related systems that have been reported and enable testing of this prodrug against cancer cell lines in vitro and in vivo.
Sigma-Aldrich
2-Thiopheneacetyl chloride, 98%
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)