MilliporeSigma
  • Carnosic acid inhibits the growth of ER-negative human breast cancer cells and synergizes with curcumin.
Carnosic acid inhibits the growth of ER-negative human breast cancer cells and synergizes with curcumin.
Fitoterapia (2012-07-26)
Linda Saxe Einbond, Hsan-Au Wu, Ryota Kashiwazaki, Kan He, Marc Roller, Tao Su, Xiaomei Wang, Sarah Goldsberry
ABSTRACT
Studies indicate that extracts and purified components, including carnosic acid, from the herb rosemary display significant growth inhibitory activity on a variety of cancers. This paper examines the ability of rosemary/carnosic acid to inhibit the growth of human breast cancer cells and to synergize with curcumin. To do this, we treated human breast cancer cells with rosemary/carnosic acid and assessed effects on cell proliferation, cell cycle distribution, gene expression patterns, activity of the purified Na/K ATPase and combinations with curcumin. Rosemary/carnosic acid potently inhibits proliferation of ER-negative human breast cancer cells and induces G1 cell cycle arrest. Further, carnosic acid is selective for MCF7 cells transfected for Her2, indicating that Her2 may function in its action. To reveal primary effects, we treated ER-negative breast cancer cells with carnosic acid for 6h. At a low dose, 5 μg/ml (15 μM), carnosic acid activated the expression of 3 genes, induced through the presence of antioxidant response elements, including genes involved in glutathione biosynthesis (CYP4F3, GCLC) and transport (SLC7A11). At a higher dose, 20 μg/ml, carnosic acid activated the expression of antioxidant (AKR1C2, TNXRD1, HMOX1) and apoptosis (GDF15, PHLDA1, DDIT3) genes and suppressed the expression of inhibitor of transcription (ID3) and cell cycle (CDKN2C) genes. Carnosic acid exhibits synergy with turmeric/curcumin. These compounds inhibited the activity of the purified Na-K-ATPase which may contribute to this synergy. Rosemary/carnosic acid, alone or combined with curcumin, may be useful to prevent and treat ER-negative breast cancer.
Sigma-Aldrich
Carnosic acid from Rosmarinus officinalis, ≥91%, powder