MilliporeSigma
  • Morphological and physiological idiosyncrasies lead to interindividual variation in flight metabolic rate in worker bumblebees (Bombus impatiens).
Morphological and physiological idiosyncrasies lead to interindividual variation in flight metabolic rate in worker bumblebees (Bombus impatiens).
Physiological and biochemical zoology : PBZ (2012-10-27)
Dimitri A Skandalis, Charles-A Darveau
ABSTRACT
Although intraspecific variation in metabolic rate is associated with variation in body size, similarly sized individuals nonetheless vary greatly. At similar masses, hovering bumblebee workers (Bombus impatiens) can differ in metabolic rate up to twofold. We examined how such interindividual variation arises by studying covariation of flight metabolic rate with morphological and other physiological parameters. Body size alone explained roughly half the variation in flight metabolic rate. The remaining variation could be explained as the outcome of variation in wing morphology and possibly an association with variation in flight muscle metabolic enzyme activities. As shown using statistical models, for a given mass, higher metabolic rate was correlated with both higher thoracic temperature and higher wing stroke frequency, in turn resulting from smaller wing surface area. When organismal and cellular metabolism for a given mass were linked, variation in metabolic rate was positively correlated with the activities of trehalase and hexokinase. Altogether, covariation with morphology and other physiological parameters explains up to 75% of the variation in metabolic rate. We also investigated the role of flight experience and show that neither flight restriction nor the number of lifetime flights affected flight energetics or flight muscle phenotype. Additionally, manipulating the level of wing asymmetry increased flight wing stroke frequency, metabolic rate, and thoracic temperature, but it did not alter enzyme activity. We conclude that idiosyncrasies in body morphology largely explained interindividual variation in flight metabolic rate but flight muscle metabolic phenotype shows little variation associated with differences in flight experience.
Sigma-Aldrich
Trehalase from porcine kidney, buffered aqueous glycerol solution, ≥1.0 units/mg protein