MilliporeSigma
  • Tannic acid and thiocarbohydrazide as structural reinforcement agents in the preparation of rabbit knee articular cartilage for the scanning electron microscope.
Tannic acid and thiocarbohydrazide as structural reinforcement agents in the preparation of rabbit knee articular cartilage for the scanning electron microscope.
The Histochemical journal (1999-07-16)
D Levanon, H Stein
ABSTRACT
Samples from seven sectors of the rabbit knee articular cartilage were shaved and prepared for the scanning electron microscope using either tannic acid, thiocarbohydrazide or nothing (control). Surface morphology was found to be more typical to a given sector and less so to a specific preparation procedure. Rough areas were recorded from load-bearing sectors, while smooth areas appeared on load-free ones. However, fibrillations were discerned on control load-bearing sectors only, and pits and humps were never detected. Tannic acid and thiocarbohydrazide may have exerted their structural reinforcing effect on the tissue preservation by enhancing the binding of osmium tetroxide to it, possibly along with that of other soluble tissue constituents.
Sigma-Aldrich
Thiocarbohydrazide, 98%