MilliporeSigma
Effect of ciprofloxacin on the pharmacokinetics of ropivacaine.
European journal of clinical pharmacology (2003-03-01)
Mika J Jokinen, Klaus T Olkkola, Jouni Ahonen, Pertti J Neuvonen
ABSTRACT
To assess the effect of ciprofloxacin on the pharmacokinetics of ropivacaine. METHODS. In a double-blind, randomised, cross-over study, nine healthy volunteers were treated for 2.5 days with 500 mg oral ciprofloxacin or placebo twice daily. On day 3, they received a single dose of 0.6 mg/kg ropivacaine intravenously over 30 min. Ropivacaine, 3-hydroxyropivacaine (3-OH-ropivacaine), and (S)-2',6'-pipecoloxylidide (PPX) in venous plasma and urine were measured for up to 12 h and 24 h, respectively. Ciprofloxacin decreased the mean clearance (CL) of ropivacaine by 31% (P<0.05), with a considerable inter-individual variation (range from -52% to +39%). It also decreased the area under the plasma concentration-time curve (AUC) of 3-OH-ropivacaine by 38% (P<0.05) and urinary excretion of 3-OH-ropivacaine by 27% (P<0.05). Ciprofloxacin increased the AUC of PPX by 71% (P<0.01) and urinary excretion of PPX by 97% (P<0.01). Ciprofloxacin modestly decreased the mean ropivacaine CL by inhibiting the CYP1A2-mediated formation of 3-OH-ropivacaine. At the same time, the CYP3A4-mediated formation of PPX was increased. There was a marked inter-individual variation in the extent of the interaction, and, for some individuals, the concomitant use of ciprofloxacin with ropivacaine might produce toxic symptoms.
Product Description
Mepivacaine impurity B, European Pharmacopoeia (EP) Reference Standard