MilliporeSigma
  • Enantioseparation of 2-aryloxypropionic acids on chiral porous monolithic columns by capillary electrochromatography. Evaluation of column performance and enantioselectivity.
Enantioseparation of 2-aryloxypropionic acids on chiral porous monolithic columns by capillary electrochromatography. Evaluation of column performance and enantioselectivity.
Journal of chromatography. A (2005-12-21)
Antonella Messina, Miroslav Flieger, Fiorella Bachechi, Massimo Sinibaldi
ABSTRACT
The enantioseparation of 2-aryloxypropionic acids by capillary electrochromatography was tested on columns with a monolithic stationary phase prepared from silanized fused-silica capillaries (100 microm I.D.) by in situ copolymerization of glycidyl methacrylate, ethylene glycol dimethacrylate and methyl methacrylate in the presence of formamide and 1-propanol as the porogen solvents. The porous chiral monolithic stationary phases were prepared by reaction of the epoxy-groups at the surface of the monolith with (+)-1-(4-aminobutyl)-(5R,8S,10R)-terguride. To attain the minimum HETP values for the enantiodiscrimination of 2-phenoxypropionic acid, the influence of the composition of polymerization solution on column total porosity and efficiency was investigated. Optimum mobile phase conditions were found for all analytes tested using acetonitrile-methanol mixtures containing triethylamine and acetic acid as the buffer components. Furthermore, the chemical and mechanical stabilities of the columns were satisfactory, allowing hundreds of analyses.
Product Description
Supelco
Haloxyfop, PESTANAL®, analytical standard