MilliporeSigma
  • Radical reactions with 3H-quinazolin-4-ones: synthesis of deoxyvasicinone, mackinazolinone, luotonin A, rutaecarpine and tryptanthrin.
Radical reactions with 3H-quinazolin-4-ones: synthesis of deoxyvasicinone, mackinazolinone, luotonin A, rutaecarpine and tryptanthrin.
Organic & biomolecular chemistry (2006-12-14)
W Russell Bowman, Mark R J Elsegood, Tobias Stein, George W Weaver
ABSTRACT
Alkyl, aryl, heteroaryl and acyl radicals have been cyclised onto the 2-position of 3H-quinazolin-4-one. The side chains containing the radical precursors were attached to the nitrogen atom in the 3-position. The cyclisations take place by aromatic homolytic substitution hence retain the aromaticity of the 3H-quinazolin-4-one ring. The highest yields were obtained using hexamethylditin to facilitate cyclisation rather than reduction without cyclisation. The alkaloids deoxyvasicinone , mackinazolinone , tryptanthrin , luotonin A and rutaecarpine were synthesised by radical cyclisation onto 3H-quinazolin-4-one.
Sigma-Aldrich
Tryptanthrin, ≥98% (HPLC)
Sigma-Aldrich
1-Chloro-4-iodobutane, 98%