MilliporeSigma
  • Pharmacological, pharmacokinetic, and primate analgesic efficacy profile of the novel bradykinin B1 Receptor antagonist ELN441958.
Pharmacological, pharmacokinetic, and primate analgesic efficacy profile of the novel bradykinin B1 Receptor antagonist ELN441958.
The Journal of pharmacology and experimental therapeutics (2007-05-02)
Jon E Hawkinson, Balazs G Szoke, Albert W Garofalo, Dennis S Hom, Hongbing Zhang, Mark Dreyer, Juri Y Fukuda, Linda Chen, Bhushan Samant, Stellanie Simmonds, Karla P Zeitz, Angie Wadsworth, Anna Liao, Raymond A Chavez, Wes Zmolek, Lany Ruslim, Michael P Bova, Ryan Holcomb, Eduardo R Butelman, Mei-Chuan Ko, Annika B Malmberg
ABSTRACT
The bradykinin B(1) receptor plays a critical role in chronic pain and inflammation, although efforts to demonstrate efficacy of receptor antagonists have been hampered by species-dependent potency differences, metabolic instability, and low oral exposure of current agents. The pharmacology, pharmacokinetics, and analgesic efficacy of the novel benzamide B(1) receptor antagonist 7-chloro-2-[3-(9-pyridin-4-yl-3,9-diazaspiro[5.5]undecanecarbonyl)phenyl]-2,3-dihydro-isoindol-1-one (ELN441958) is described. ELN441958 competitively inhibited the binding of the B(1) agonist ligand [(3)H]desArg(10)-kallidin ([(3)H]DAKD) to IMR-90 human fibroblast membranes with high affinity (K(i) = 0.26 +/- 0.02 nM). ELN441958 potently antagonized DAKD (but not bradykinin)-induced calcium mobilization in IMR-90 cells, indicating that it is highly selective for B(1) over B(2) receptors. Antagonism of agonist-induced calcium responses at B(1) receptors from different species indicated that ELN441958 is selective for primate over rodent B(1) receptors with a rank order potency (K(B), nanomolar) of human (0.12 +/- 0.02) approximately rhesus monkey (0.24 +/- 0.01) > rat (1.5 +/- 0.4) > mouse (14 +/- 4). ELN441958 had good permeability and metabolic stability in vitro consistent with high oral exposure and moderate plasma half-lives in rats and rhesus monkeys. Because ELN441958 is up to 120-fold more potent at primate than at rodent B(1) receptors, it was evaluated in a primate pain model. ELN441958 dose-dependently reduced carrageenan-induced thermal hyperalgesia in a rhesus monkey tail-withdrawal model, with an ED(50) approximately 3 mg/kg s.c. Naltrexone had no effect on the antihyperalgesia produced by ELN441958, indicating a lack of involvement of opioid receptors. ELN441958 is a novel small molecule bradykinin B(1) receptor antagonist exhibiting high oral bioavailability and potent systemic efficacy in rhesus monkey inflammatory pain.
Sigma-Aldrich
[Lys-des-Arg9]-Bradykinin, ≥95% (HPLC)