MilliporeSigma
  • Lipid classes and fatty acid composition of the tropical nudibranch mollusks Chromodoris sp. and Phyllidia coelestis.
Lipid classes and fatty acid composition of the tropical nudibranch mollusks Chromodoris sp. and Phyllidia coelestis.
Lipids (2007-10-26)
Natalia V Zhukova
ABSTRACT
Two nudibranch mollusks, Chromodoris sp. and Phyllidia coelestis, collected from tropical waters of the Northwestern Pacific, were analyzed for lipids. The aim of this study was to fill the gap in knowledge of lipid biochemistry of mollusks. Phospholipids (PL) were the dominating lipid class followed by sterols (13%). Neutral lipids were not detected in Chromodoris sp. By contrast, P. coelestis contained TAG, diacylglyceryl ether, long chain alcohol and esters of sterols. Among PL, PC was predominant (about 50%); PE, PS and CAEP were almost in equal proportions. Sixty five FA were identified as methyl esters and N-acyl pyrrolidides by GC-MS. The sea slugs exhibited a wide diversity of FA. The common marine n-3 PUFA, 20:5n-6 and 22:6n-3, constituted 0.6-1.3% of the total FA, whereas n-6 PUFA, 22:4n-6, 20:4n-6, and 18:2n-6, were the main (25%). Among monounsaturated FA, 7-21:1 was the main (up to 6.2%). The non-methylene-interrupted (NMI) FA were found (9.4 and 12.4%), including the known 5,11-20:2, 5,13-20:2, 7,13-22:2, 7,15-22:2 and a novel isomer 7,13-21:2 (up to 3.9%). The pathway of its biosynthesis was suggested. A series of very long chain FA (VLC FA), with the main 5,9-25:2 and 5,9-26:2, were identified. High level of VLC FA (8.7 and 11.7%) in sea slugs is apparently the result of predation on sponges. Another unique feature concerned a high abundance of various odd and branched FA (16.7 and 34%), which could have originated from the dietary origin or symbiotic bacteria. This is the first report on lipid and FA composition of nudibranchs.
Sigma-Aldrich
2-Aminoethylphosphonic acid, 99%