MilliporeSigma
  • A meta effect in nonphotochemical processes: the homolytic chemistry of m-methoxyphenol.
A meta effect in nonphotochemical processes: the homolytic chemistry of m-methoxyphenol.
The Journal of organic chemistry (2008-02-26)
Mario C Foti, Carmelo Daquino, Gino A DiLabio, K U Ingold
ABSTRACT
The m-methoxy group is normally electron-withdrawing (EW), sigma(m) = +0.12, sigma(m+) = +0.05. The strong EW activity of a phenoxyl radical's O* atom causes the m-methoxy group to become electron-donating (ED), sigma(m)(+) = -0.14. In valence bond terms, this can be ascribed to the nonclassical resonance structures 1c-e. Although it has long been known that m-methoxy is ED in photoexcited states, it has now been found to be ED for homolytic O-H bond breaking in ground-state 3-methoxyphenol.
Sigma-Aldrich
3-Methoxyphenol, 96%